Assay Details
| Gene Symbol: CG9389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141038.3 | NP_649295.1 | 2 - 3 | 1538 | 78 | |
| GenBank mRNA | AY060713.1 | - | 2 - 3 | 1489 | 78 | |
More Information
Set Membership: |
|

CG9389
Genomic Map