Assay Details
| Gene Symbol: Alp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141058.3 | NP_649315.1 | 3 - 4 | 1515 | 66 | |
| GenBank mRNA | AY069739.1 | - | 3 - 4 | 1499 | 66 | |
More Information
Set Membership: |
|

Alp1
Genomic Map