Assay Details
| Gene Symbol: SAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141067.3 | NP_649324.1 | 5 - 6 | 2012 | 75 | |
| GenBank mRNA | AF106952.1 | - | 5 - 6 | 1936 | 75 | |
| BT044561.1 | - | 5 - 6 | 2009 | 75 | ||
More Information
Set Membership: |
|

SAK
Genomic Map