Assay Details
| Gene Symbol: SAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141067.3 | NP_649324.1 | 6 - 7 | 2161 | 109 | |
| GenBank mRNA | AF106952.1 | - | 6 - 7 | 2085 | 109 | |
| BT044561.1 | - | 6 - 7 | 2158 | 109 | ||
More Information
Set Membership: |
|

SAK
Genomic Map