Assay Details
| Gene Symbol: Cdk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275238.1 | NP_001262167.1 | 2 - 3 | 1457 | 63 | |
| NM_141068.4 | NP_649325.2 | 2 - 3 | 1457 | 63 | ||
| NM_168912.2 | NP_730643.1 | 3 - 4 | 1743 | 63 | ||
| GenBank mRNA | BT058001.1 | - | 2 - 3 | 1392 | 63 | |
More Information
Set Membership: |
|

Cdk12
Genomic Map