Assay Details
| Gene Symbol: CG7611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170018.2 | NP_001163489.1 | 1 - 2 | 2533 | 74 | |
| NM_141069.4 | NP_649326.1 | 2 - 3 | 1879 | 74 | ||
| NM_168913.3 | NP_730644.1 | 2 - 3 | 1417 | 74 | ||
| NM_168914.3 | NP_730645.1 | 2 - 3 | 1139 | 74 | ||
| NM_168915.3 | NP_730646.1 | 2 - 3 | 1212 | 74 | ||
| NM_168916.3 | NP_730647.1 | 2 - 3 | 1041 | 74 | ||
| NM_168917.3 | NP_730648.1 | 2 - 3 | 1070 | 74 | ||
| NM_168918.3 | NP_730649.1 | 2 - 3 | 1439 | 74 | ||
| NM_168919.3 | NP_730650.1 | 2 - 3 | 1110 | 74 | ||
| GenBank mRNA | AY069717.1 | - | 1 - 2 | 2436 | 74 | |
More Information
Set Membership: |
|

CG7611
Genomic Map