Assay Details
| Gene Symbol: TfAP-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275248.1 | NP_001262177.1 | 2 - 3 | 295 | 97 | |
| NM_141079.3 | NP_649336.1 | 2 - 3 | 709 | 97 | ||
| NM_168927.2 | NP_730664.1 | 2 - 3 | 484 | 97 | ||
| GenBank mRNA | AF033030.1 | - | 2 - 3 | 693 | 97 | |
| AJ006868.1 | - | 2 - 3 | 484 | 97 | ||
| AY071140.1 | - | 2 - 3 | 707 | 97 | ||
More Information
Set Membership: |
|

TfAP-2
Genomic Map