Assay Details
| Gene Symbol: TfAP-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275248.1 | NP_001262177.1 | 4 - 5 | 683 | 60 | |
| NM_001275249.1 | NP_001262178.1 | 4 - 5 | 1115 | 60 | ||
| NM_001275250.1 | NP_001262179.1 | 4 - 5 | 890 | 60 | ||
| NM_001275251.1 | NP_001262180.1 | 4 - 5 | 701 | 60 | ||
| NM_141079.3 | NP_649336.1 | 4 - 5 | 1097 | 60 | ||
| NM_168927.2 | NP_730664.1 | 4 - 5 | 872 | 60 | ||
| GenBank mRNA | AF033030.1 | - | 4 - 5 | 1081 | 60 | |
| AJ006868.1 | - | 4 - 5 | 872 | 60 | ||
| AY071140.1 | - | 4 - 5 | 1095 | 60 | ||
More Information
Set Membership: |
|

TfAP-2
Genomic Map