Assay Details
| Gene Symbol: TfAP-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275248.1 | NP_001262177.1 | 6 - 7 | 1127 | 66 | |
| NM_001275249.1 | NP_001262178.1 | 6 - 7 | 1559 | 66 | ||
| NM_001275250.1 | NP_001262179.1 | 6 - 7 | 1334 | 66 | ||
| NM_001275251.1 | NP_001262180.1 | 6 - 7 | 1145 | 66 | ||
| NM_141079.3 | NP_649336.1 | 6 - 7 | 1541 | 66 | ||
| NM_168927.2 | NP_730664.1 | 6 - 7 | 1316 | 66 | ||
| GenBank mRNA | AF033030.1 | - | 6 - 7 | 1525 | 66 | |
| AJ006868.1 | - | 6 - 7 | 1316 | 66 | ||
| AY071140.1 | - | 6 - 7 | 1539 | 66 | ||
More Information
Set Membership: |
|

TfAP-2
Genomic Map