Assay Details
| Gene Symbol: S1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141080.3 | NP_649337.2 | 1 - 2 | 230 | 116 | |
| GenBank mRNA | AF441758.1 | - | 1 - 2 | 218 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

S1P| Gene Symbol: S1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141080.3 | NP_649337.2 | 1 - 2 | 230 | 116 | |
| GenBank mRNA | AF441758.1 | - | 1 - 2 | 218 | 116 | |
Set Membership: |
|