Assay Details
| Gene Symbol: S1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141080.3 | NP_649337.2 | 3 - 4 | 1620 | 110 | |
| GenBank mRNA | AF441758.1 | - | 3 - 4 | 1608 | 110 | |
More Information
Set Membership: |
|

S1P
Genomic Map