Assay Details
| Gene Symbol: S1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141080.3 | NP_649337.2 | 5 - 6 | 2917 | 100 | |
| GenBank mRNA | AF441758.1 | - | 5 - 6 | 2905 | 100 | |
More Information
Set Membership: |
|

S1P
Genomic Map