Assay Details
| Gene Symbol: CG7414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144524.3 | NP_001137996.2 | 2 - 3 | 1276 | 86 | |
| NM_001202226.1 | NP_001189155.1 | 2 - 3 | 1501 | 86 | ||
| NM_001202227.2 | NP_001189156.1 | 2 - 3 | 1276 | 86 | ||
| NM_141108.2 | NP_649365.1 | 2 - 3 | 1501 | 86 | ||
| GenBank mRNA | AY051717.1 | - | 2 - 3 | 1275 | 86 | |
More Information
Set Membership: |
|

CG7414
Genomic Map