Assay Details
| Gene Symbol: CG7414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144524.3 | NP_001137996.2 | 3 - 4 | 1557 | 76 | |
| NM_141108.2 | NP_649365.1 | 3 - 4 | 1782 | 76 | ||
| GenBank mRNA | AY051717.1 | - | 3 - 4 | 1556 | 76 | |
More Information
Set Membership: |
|

CG7414
Genomic Map