Assay Details
| Gene Symbol: Srpk79D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202231.2 | NP_001189160.1 | 2 - 3 | 1007 | 80 | |
| NM_001259950.2 | NP_001246879.1 | 1 - 2 | 1018 | 80 | ||
| NM_001259953.2 | NP_001246882.1 | 1 - 2 | 1018 | 80 | ||
| NM_001275279.1 | NP_001262208.1 | 1 - 2 | 1356 | 80 | ||
| NM_001275280.1 | NP_001262209.1 | 1 - 2 | 1018 | 80 | ||
| NM_001275281.1 | NP_001262210.1 | 1 - 2 | 1018 | 80 | ||
| NM_141130.4 | NP_649387.2 | 2 - 3 | 1007 | 80 | ||
| GenBank mRNA | AY094630.1 | - | 2 - 3 | 1007 | 80 | |
More Information
Set Membership: |
|

Srpk79D
Genomic Map