Assay Details
| Gene Symbol: Smyd4-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165831.3 | NP_725048.1 | 2 - 3 | 478 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smyd4-3| Gene Symbol: Smyd4-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165831.3 | NP_725048.1 | 2 - 3 | 478 | 117 | |
Set Membership: |
|