Assay Details
| Gene Symbol: E(Pc) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273966.2 | NP_001260895.1 | 9 - 10 | 5745 | 59 | |
| NM_001273967.1 | NP_001260896.1 | 10 - 11 | 6116 | 59 | ||
| NM_001273968.1 | NP_001260897.1 | 10 - 11 | 6116 | 59 | ||
| NM_078974.3 | NP_523698.2 | 10 - 11 | 5970 | 59 | ||
| NM_165836.3 | NP_725054.1 | 10 - 11 | 5887 | 59 | ||
More Information
Set Membership: |
|

E(Pc)
Genomic Map