Assay Details
| Gene Symbol: E(Pc) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273966.2 | NP_001260895.1 | 2 - 3 | 1198 | 70 | |
| NM_001273967.1 | NP_001260896.1 | 2 - 3 | 1422 | 70 | ||
| NM_001273968.1 | NP_001260897.1 | 2 - 3 | 1422 | 70 | ||
| NM_078974.3 | NP_523698.2 | 2 - 3 | 1276 | 70 | ||
| NM_165836.3 | NP_725054.1 | 2 - 3 | 1193 | 70 | ||
More Information
Set Membership: |
|

E(Pc)
Genomic Map