Assay Details
| Gene Symbol: tou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103800.3 | NP_001097270.1 | 15 - 16 | 9640 | 85 | |
| NM_001273969.1 | NP_001260898.1 | 15 - 16 | 9122 | 85 | ||
| NM_001273970.1 | NP_001260899.1 | 14 - 15 | 9495 | 85 | ||
| NM_078977.5 | NP_523701.3 | 15 - 16 | 9244 | 85 | ||
| NM_165844.2 | NP_725062.2 | 3 - 4 | 1702 | 85 | ||
| GenBank mRNA | AY060335.1 | - | 3 - 4 | 1702 | 85 | |
More Information
Set Membership: |
|

tou
Genomic Map