Assay Details
| Gene Symbol: Cam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259347.2 | NP_001246276.1 | 3 - 4 | 382 | 70 | |
| NM_001259348.3 | NP_001246277.1 | 2 - 3 | 304 | 70 | ||
| NM_001299408.1 | NP_001286337.1 | 2 - 3 | 329 | 70 | ||
| NM_078986.3 | NP_523710.1 | 3 - 4 | 382 | 70 | ||
| NM_165870.2 | NP_725120.1 | 2 - 3 | 304 | 70 | ||
| GenBank mRNA | AY118890.1 | - | 3 - 4 | 359 | 70 | |
| BT003282.1 | - | 2 - 3 | 424 | 70 | ||
More Information
Set Membership: |
|

Cam
Genomic Map