Assay Details
| Gene Symbol: garz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259349.2 | NP_001246278.1 | 4 - 5 | 2579 | 69 | |
| NM_136917.4 | NP_610761.2 | 4 - 5 | 2579 | 69 | ||
| NM_165881.2 | NP_725133.1 | 4 - 5 | 2579 | 69 | ||
More Information
Set Membership: |
|

garz
Genomic Map