Assay Details
| Gene Symbol: garz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259349.2 | NP_001246278.1 | 6 - 7 | 3013 | 60 | |
| NM_136917.4 | NP_610761.2 | 6 - 7 | 3013 | 60 | ||
| NM_165881.2 | NP_725133.1 | 6 - 7 | 3013 | 60 | ||
More Information
Set Membership: |
|

garz
Genomic Map