Assay Details
Gene Symbol: Cpr49Ac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165892.4 | NP_725150.2 | 2 - 3 | 268 | 70 | |
GenBank mRNA | BT003229.1 | - | 2 - 3 | 249 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
