Assay Details
| Gene Symbol: fra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299424.1 | NP_001286353.1 | 3 - 4 | 2608 | 86 | |
| NM_078992.3 | NP_523716.2 | 3 - 4 | 2635 | 86 | ||
| NM_165913.2 | NP_725185.1 | 3 - 4 | 2182 | 86 | ||
| GenBank mRNA | BT150366.1 | - | 3 - 4 | 2632 | 86 | |
More Information
Set Membership: |
|

fra
Genomic Map