Assay Details
| Gene Symbol: Galphaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165919.2 | NP_725191.1 | 3 - 4 | 1434 | 83 | |
| NM_165920.3 | NP_725192.2 | 2 - 3 | 622 | 83 | ||
| NM_165921.2 | NP_725193.1 | 2 - 3 | 622 | 83 | ||
| NM_165922.2 | NP_725194.1 | 3 - 4 | 471 | 83 | ||
| NM_165923.2 | NP_725195.1 | 3 - 4 | 1401 | 83 | ||
| NM_165924.2 | NP_725196.1 | 2 - 3 | 553 | 83 | ||
| NM_165925.3 | NP_725197.2 | 2 - 3 | 655 | 83 | ||
| GenBank mRNA | AY118511.1 | - | 3 - 4 | 1402 | 83 | |
| BT024269.1 | - | 1 - 2 | 501 | 83 | ||
| U31092.1 | - | 3 - 4 | 1429 | 83 | ||
| Gene Symbol: CG45086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299427.1 | NP_001286356.1 | 3 - 4 | 1401 | 83 | |
| NM_001299428.1 | NP_001286357.1 | 3 - 4 | 1434 | 83 | ||
More Information
Set Membership: |
|

Galphaq
Genomic Map