Assay Details
| Gene Symbol: Galphaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165919.2 | NP_725191.1 | 7 - 8 | 2032 | 117 | |
| NM_165920.3 | NP_725192.2 | 7 - 8 | 1349 | 117 | ||
| NM_165921.2 | NP_725193.1 | 6 - 7 | 1220 | 117 | ||
| NM_165922.2 | NP_725194.1 | 7 - 8 | 1069 | 117 | ||
| NM_165923.2 | NP_725195.1 | 7 - 8 | 1999 | 117 | ||
| NM_165924.2 | NP_725196.1 | 6 - 7 | 1151 | 117 | ||
| NM_165925.3 | NP_725197.2 | 6 - 7 | 1253 | 117 | ||
| GenBank mRNA | AY118511.1 | - | 7 - 8 | 2000 | 117 | |
| BT024269.1 | - | 5 - 6 | 1099 | 117 | ||
| U31092.1 | - | 7 - 8 | 2027 | 117 | ||
| Gene Symbol: CG45086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299427.1 | NP_001286356.1 | 7 - 8 | 1999 | 117 | |
| NM_001299428.1 | NP_001286357.1 | 7 - 8 | 2032 | 117 | ||
More Information
Set Membership: |
|

Galphaq
Genomic Map