Assay Details
| Gene Symbol: Galphaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165919.2 | NP_725191.1 | 9 - 10 | 2290 | 122 | |
| NM_165921.2 | NP_725193.1 | 8 - 9 | 1478 | 122 | ||
| NM_165922.2 | NP_725194.1 | 9 - 10 | 1327 | 122 | ||
| NM_165923.2 | NP_725195.1 | 9 - 10 | 2257 | 122 | ||
| NM_165925.3 | NP_725197.2 | 8 - 9 | 1511 | 122 | ||
| GenBank mRNA | AY118511.1 | - | 9 - 10 | 2258 | 122 | |
| U31092.1 | - | 9 - 10 | 2285 | 122 | ||
| Gene Symbol: CG45086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299427.1 | NP_001286356.1 | 9 - 10 | 2257 | 122 | |
| NM_001299428.1 | NP_001286357.1 | 9 - 10 | 2290 | 122 | ||
More Information
Set Membership: |
|

Galphaq
Genomic Map