Assay Details
| Gene Symbol: Galphaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165924.2 | NP_725196.1 | 7 - 8 | 1306 | 133 | |
| GenBank mRNA | BT024269.1 | - | 6 - 7 | 1254 | 133 | |
More Information
Set Membership: |
|

Galphaq
Genomic Map