Assay Details
| Gene Symbol: CG30054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043073.3 | NP_001036538.3 | 4 - 5 | 585 | 96 | |
| NM_001201985.1 | NP_001188914.1 | 3 - 4 | 590 | 96 | ||
More Information
Set Membership: |
|

CG30054
Genomic Map