Assay Details
| Gene Symbol: CG30054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043073.3 | NP_001036538.3 | 6 - 7 | 875 | 98 | |
| NM_001201985.1 | NP_001188914.1 | 5 - 6 | 880 | 98 | ||
More Information
Set Membership: |
|

CG30054
Genomic Map