Assay Details
| Gene Symbol: Iswi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078995.2 | NP_523719.1 | 4 - 5 | 2909 | 145 | |
| NM_165930.2 | NP_725203.1 | 5 - 6 | 2803 | 145 | ||
| NM_165931.2 | NP_725204.1 | 5 - 6 | 2847 | 145 | ||
| GenBank mRNA | BT044562.1 | - | 5 - 6 | 2800 | 145 | |
| L27127.1 | - | 4 - 5 | 2962 | 145 | ||
More Information
Set Membership: |
|

Iswi
Genomic Map