Assay Details
| Gene Symbol: sca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057362.3 | NP_476710.2 | 3 - 4 | 1781 | 60 | |
| NM_165951.2 | NP_725230.2 | 3 - 4 | 1781 | 60 | ||
| GenBank mRNA | AY129456.1 | - | 3 - 4 | 1778 | 60 | |
More Information
Set Membership: |
|

sca
Genomic Map