Assay Details
| Gene Symbol: TppII | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165966.2 | NP_725252.1 | 1 - 2 | 376 | 66 | |
| GenBank mRNA | BT044288.1 | - | 1 - 2 | 327 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TppII| Gene Symbol: TppII | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165966.2 | NP_725252.1 | 1 - 2 | 376 | 66 | |
| GenBank mRNA | BT044288.1 | - | 1 - 2 | 327 | 66 | |
Set Membership: |
|