Assay Details
| Gene Symbol: TppII | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299447.1 | NP_001286376.1 | 2 - 3 | 369 | 103 | |
| NM_001299448.1 | NP_001286377.1 | 1 - 2 | 281 | 103 | ||
| NM_057899.3 | NP_477247.1 | 1 - 2 | 281 | 103 | ||
| NM_165966.2 | NP_725252.1 | 2 - 3 | 635 | 103 | ||
| GenBank mRNA | BT003260.1 | - | 1 - 2 | 273 | 103 | |
| BT044288.1 | - | 2 - 3 | 586 | 103 | ||
More Information
Set Membership: |
|

TppII
Genomic Map