Assay Details
| Gene Symbol: TppII | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299447.1 | NP_001286376.1 | 6 - 7 | 1932 | 115 | |
| NM_001299448.1 | NP_001286377.1 | 6 - 7 | 1907 | 115 | ||
| NM_057899.3 | NP_477247.1 | 5 - 6 | 1844 | 115 | ||
| NM_165966.2 | NP_725252.1 | 6 - 7 | 2198 | 115 | ||
| GenBank mRNA | BT003260.1 | - | 5 - 6 | 1836 | 115 | |
| BT044288.1 | - | 6 - 7 | 2149 | 115 | ||
More Information
Set Membership: |
|

TppII
Genomic Map