Assay Details
| Gene Symbol: ValRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274017.1 | NP_001260946.1 | 2 - 3 | 291 | 58 | |
| NM_080099.3 | NP_524838.1 | 1 - 2 | 242 | 58 | ||
| NM_165970.2 | NP_725259.1 | 2 - 3 | 278 | 58 | ||
More Information
Set Membership: |
|

ValRS
Genomic Map