Assay Details
| Gene Symbol: Dp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274020.1 | NP_001260949.1 | 6 - 7 | 981 | 79 | |
| NM_057691.4 | NP_477039.1 | 6 - 7 | 1594 | 79 | ||
| NM_165974.3 | NP_725267.1 | 5 - 6 | 1059 | 79 | ||
| GenBank mRNA | AY069526.1 | - | 6 - 7 | 1543 | 79 | |
| BT056266.1 | - | 5 - 6 | 1057 | 79 | ||
More Information
Set Membership: |
|

Dp
Genomic Map