Assay Details
| Gene Symbol: cnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038855.3 | NP_001033944.1 | 2 - 3 | 599 | 89 | |
| NM_001259365.2 | NP_001246294.1 | 2 - 3 | 377 | 89 | ||
| NM_001259366.2 | NP_001246295.1 | 2 - 3 | 288 | 89 | ||
| NM_165990.3 | NP_725297.1 | 2 - 3 | 599 | 89 | ||
More Information
Set Membership: |
|

cnn
Genomic Map