Assay Details
| Gene Symbol: cnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014523.2 | NP_001014523.1 | 5 - 6 | 3125 | 58 | |
| NM_001038855.3 | NP_001033944.1 | 7 - 8 | 3636 | 58 | ||
| NM_001259364.2 | NP_001246293.1 | 10 - 11 | 3997 | 58 | ||
| NM_001259365.2 | NP_001246294.1 | 7 - 8 | 3384 | 58 | ||
| NM_001259366.2 | NP_001246295.1 | 7 - 8 | 3295 | 58 | ||
| NM_057736.6 | NP_477084.1 | 6 - 7 | 3307 | 58 | ||
| NM_165990.3 | NP_725297.1 | 7 - 8 | 3606 | 58 | ||
| NM_165991.4 | NP_725298.1 | 9 - 10 | 3815 | 58 | ||
| GenBank mRNA | BT010053.1 | - | 6 - 7 | 3277 | 58 | |
More Information
Set Membership: |
|

cnn
Genomic Map