Assay Details
| Gene Symbol: bbc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259371.2 | NP_001246300.1 | 1 - 2 | 616 | 77 | |
| NM_001274026.1 | NP_001260955.1 | 1 - 2 | 616 | 77 | ||
| NM_137033.4 | NP_610877.2 | 1 - 2 | 616 | 77 | ||
| NM_165993.3 | NP_725301.1 | 1 - 2 | 616 | 77 | ||
More Information
Set Membership: |
|

bbc
Genomic Map