Assay Details
| Gene Symbol: Cad74A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259885.1 | NP_001246814.1 | 2 - 3 | 770 | 60 | |
| NM_140716.2 | NP_648973.1 | 2 - 3 | 484 | 60 | ||
| NM_168724.2 | NP_730261.1 | 2 - 3 | 499 | 60 | ||
| GenBank mRNA | BT150474.1 | - | 2 - 3 | 498 | 60 | |
More Information
Set Membership: |
|

Cad74A
Genomic Map