Assay Details
| Gene Symbol: ftz-f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259895.2 | NP_001246824.1 | 3 - 4 | 3007 | 76 | |
| NM_079419.3 | NP_524143.2 | 5 - 6 | 3646 | 76 | ||
| NM_168775.2 | NP_730359.1 | 3 - 4 | 2219 | 76 | ||
| GenBank mRNA | BT058016.1 | - | 5 - 6 | 3232 | 76 | |
More Information
Set Membership: |
|

ftz-f1
Genomic Map