Assay Details
| Gene Symbol: ftz-f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259895.2 | NP_001246824.1 | 5 - 6 | 3262 | 85 | |
| NM_001316437.1 | NP_001303366.1 | 5 - 6 | 3412 | 85 | ||
| NM_079419.3 | NP_524143.2 | 7 - 8 | 3901 | 85 | ||
| NM_168775.2 | NP_730359.1 | 5 - 6 | 2474 | 85 | ||
| GenBank mRNA | BT058016.1 | - | 7 - 8 | 3487 | 85 | |
More Information
Set Membership: |
|

ftz-f1
Genomic Map