Assay Details
| Gene Symbol: Indy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169994.2 | NP_001163465.1 | 2 - 3 | 280 | 86 | |
| NM_079426.4 | NP_524150.1 | 2 - 3 | 183 | 86 | ||
| NM_168778.2 | NP_730363.2 | 2 - 3 | 271 | 86 | ||
| NM_168779.2 | NP_730364.1 | 2 - 3 | 299 | 86 | ||
| GenBank mRNA | AF509505.1 | - | 2 - 3 | 266 | 86 | |
| AY102686.1 | - | 2 - 3 | 147 | 86 | ||
| BT100230.1 | - | 2 - 3 | 265 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY102686.1, NM_001169994.2, NM_079426.4, NM_168779.2 transcripts.
Set Membership: |
|

Indy
Genomic Map