Assay Details
| Gene Symbol: CG3961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140810.4 | NP_649067.2 | 3 - 4 | 424 | 89 | |
| NM_168781.3 | NP_730369.1 | 2 - 3 | 488 | 89 | ||
| GenBank mRNA | BT100231.1 | - | 2 - 3 | 471 | 89 | |
More Information
Set Membership: |
|

CG3961
Genomic Map