Assay Details
| Gene Symbol: CG3961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275095.1 | NP_001262024.1 | 4 - 5 | 466 | 117 | |
| NM_140810.4 | NP_649067.2 | 5 - 6 | 753 | 117 | ||
| NM_168781.3 | NP_730369.1 | 4 - 5 | 817 | 117 | ||
| NM_168782.2 | NP_730370.2 | 3 - 4 | 496 | 117 | ||
| GenBank mRNA | BT100231.1 | - | 4 - 5 | 800 | 117 | |
| BT132918.1 | - | 3 - 4 | 496 | 117 | ||
More Information
Set Membership: |
|

CG3961
Genomic Map