Assay Details
| Gene Symbol: CG3961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275095.1 | NP_001262024.1 | 7 - 8 | 2051 | 65 | |
| NM_140810.4 | NP_649067.2 | 8 - 9 | 2338 | 65 | ||
| NM_168781.3 | NP_730369.1 | 7 - 8 | 2402 | 65 | ||
| NM_168782.2 | NP_730370.2 | 6 - 7 | 2081 | 65 | ||
| GenBank mRNA | BT100231.1 | - | 7 - 8 | 2385 | 65 | |
| BT132918.1 | - | 6 - 7 | 2081 | 65 | ||
More Information
Set Membership: |
|

CG3961
Genomic Map