Assay Details
| Gene Symbol: nes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079433.3 | NP_524157.2 | 2 - 3 | 263 | 95 | |
| NM_168788.3 | NP_730390.1 | 2 - 3 | 150 | 95 | ||
| NM_176349.2 | NP_788527.1 | 2 - 3 | 247 | 95 | ||
| GenBank mRNA | AY070936.1 | - | 2 - 3 | 146 | 95 | |
More Information
Set Membership: |
|

nes
Genomic Map