Assay Details
| Gene Symbol: Rcd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140868.2 | NP_649125.1 | 2 - 3 | 941 | 105 | |
| NM_168803.2 | NP_730429.2 | 3 - 4 | 1302 | 105 | ||
| GenBank mRNA | BT044243.1 | - | 3 - 4 | 1302 | 105 | |
More Information
Set Membership: |
|

Rcd7
Genomic Map