Assay Details
| Gene Symbol: SREBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275135.1 | NP_001262064.1 | 5 - 6 | 3012 | 88 | |
| NM_079442.3 | NP_524166.2 | 4 - 5 | 2910 | 88 | ||
| NM_168815.2 | NP_730449.1 | 5 - 6 | 2933 | 88 | ||
| NM_168816.2 | NP_730450.1 | 5 - 6 | 3043 | 88 | ||
| GenBank mRNA | BT010285.1 | - | 5 - 6 | 2924 | 88 | |
More Information
Set Membership: |
|

SREBP
Genomic Map